missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Antibody (4E1G2), Janelia Fluor™ 646 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11603JF646
This item is not returnable.
View return policy
Description
Testosterone Monoclonal antibody specifically detects Testosterone in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Testosterone | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone | |
| Testosterone 3 CMO conjugated to BSA | |
| 0.1 mL | |
| Cellular Signaling, Developmental Biology | |
| Non-species specific | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4E1G2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction