missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Antibody (14P2C8), R&D Systems™

Product Code. 18750914 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18750914 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18750914 Supplier Novus Biologicals Supplier No. NBP178562
  • ONLINE EXCLUSIVE PRICE
    522.00€
    493.50€ / 100µg
    Save 28.50€
    5% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Testosterone Monoclonal specifically detects Testosterone in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA).
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA)
Dilution ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
Purification Method Ion exchange chromatography
Target Species Non-species specific
Product Type Antibody
Product Line Novus

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.