Learn More
Terpinolene 1000 μg/mL in Isopropanol, Dr. Ehrenstorfer
Product Code. 30319706
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 30319706
Brand: Dr. Ehrenstorfer DREGA09010042IP
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Terpinolene
CAS 586-62-9
Analyte or Component Names Terpinolene
Shipping Condition Ice pack (-20°C)
Molecular Formula C10 H16
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Cyclohexene, 1-methyl-4-(1-methylethylidene)-, p-Mentha-1,4(8)-diene (8CI), 1-Methyl-4-(1-methylethylidene)cyclohexene, 4-Isopropylidene-1-methylcyclohexene, Isoterpinene, Nofmer TP, Terpinolen, Terpinolene, α-Terpinolene, Δ-Terpinene
InChI Formula InChI=1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4H,5-7H2,1-3H3
SMILES CC(=C1CCC(=CC1)C)C
IUPAC Name 1-methyl-4-propan-2-ylidenecyclohexene
Solvent or Matrix Isopropanol
Molecular Weight (g/mol) 136.23
Quantity 1 mL
Formula Weight 136.1252 g/mol