Learn More
TER119 Rat, anti-Mouse, FITC, Clone: TER-119, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 100-0454
Encompass_Preferred
 View more versions of this product
Product Code. 17117621
  • 272.70€ / 100µg
Description
  • The TER-119 antibody reacts with murine TER119 (Ly-76), an ∼52 kDa protein associated with glycophorin A on the surface of cells of the erythroid lineage in embryonic yolk sac, fetal and newborn liver, adult bone marrow, peripheral blood, and lymphoid organs
  • TER119 is an erythroid-specific marker expressed at all stages of differentiation from early proerythroblasts to mature erythrocytes, but not by erythroid colony-forming cells (BFU-E, blast-forming unit erythroid, or CFU-E, colony-forming unit erythroid)
  • The TER-119 antibody is a component of the 'lineage cocktail' used to detect, or deplete cells committed to hematopoietic lineages
  • In adult mice, TER119 is found on ∼20 - 25% of bone marrow cells and ∼2 - 3% of splenocytes
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD4+ T Cell Isolation Kit (Catalog #19852)
Specifications
Specifications
TER119
Monoclonal
FITC
100 μg
Mouse
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
TER-119
Rat
Primary
Erythroid Cells, Red Blood Cells