missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teplizumab Antibody (2505B), PE/Atto594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10245PEATT594
This item is not returnable.
View return policy
Description
Teplizumab Monoclonal antibody specifically detects Teplizumab in Broad Species Reactivity samples. It is validated for Flow Cytometry
Specifications
| Teplizumab | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Full-length Teplizumab | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
| Flow Cytometry | |
| 2505B | |
| Flow Cytometry | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction