Learn More
Buerkle™ Telescopic Rod
Product Code. 10396242 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
115 to 300 cm (Adjustable)
125 to 250 cm (Adjustable)
165 to 450 cm (Adjustable)
175 to 600 cm (Adjustable)
65 to 120 cm (Adjustable)
70 to 180 cm
95 to 280 cm (Adjustable)
Includes:
2 Rods
3 Rods
4 Rods
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Includes unitSize
10396242 65 to 120 cm (Adjustable) 2 Rods Each
10530053 125 to 250 cm (Adjustable) 2 Rods Each
15376297 70 to 180 cm 4 Rods Each
10376292 95 to 280 cm (Adjustable) 4 Rods Each
10040822 115 to 300 cm (Adjustable) 3 Rods Each
10182412 165 to 450 cm (Adjustable) 3 Rods Each
10244992 175 to 600 cm (Adjustable) 4 Rods Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10396242 Supplier Buerkle™ Supplier No. 53550120
In Stock
Add to basket
Add to basket
Versatile sampling system with exchangeable tools for a wide range of applications.
  • Different tools can be attached to the telescopic rod
  • The tools (angular beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net) are attached to the telescopic rod by a practical snap-in joint
  • Ideal for sampling from baths, tanks, shafts, open water courses etc
  • Sampling depths down to 6 m can be reached with the adjustable telescopic aluminum rod
  • The telescopic rods cannot be rotated radially, which means even better handling
  • The individual rods have been provided with an end stop, this means that one rod cannot accidentally slip out of another rod
  • Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm. Fits into every car boot!
  • Springs made of stainless steel
TRUSTED_SUSTAINABILITY
Specifications
Product Type Telescopic Rod
Material Aluminum
Autoclavable Not Autoclavable
Includes 2 Rods
For Use With (Application) Industry and Water Treatment, Ideal for Sampling From Baths, Tanks, Shafts, Open Water Courses, Used with Telescope Sampler
For Use With (Equipment) Bürkle Angled beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net
Length (Metric) 65 to 120 cm (Adjustable)