missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdT Mouse anti-Human, Clone: TDT/1393, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | TdT |
|---|---|
| Clone | TDT/1393 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | ELISA, Immunohistochemistry (Paraffin), Peptide Array |
Description
TdT Monoclonal specifically detects TdT in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
| TdT | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 10 mM PBS with 0.05% BSA | |
| 1791 | |
| IgG1 κ | |
| Protein A or G purified |
| TDT/1393 | |
| ELISA 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase | |
| Recombinant fragment (around aa 52-192) of human TdT protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title