Learn More
TDS Calibration Standard 1382ppm, Jenway™
Product Code. 11490579
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490579 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11490579 Supplier Jenway Supplier No. 1615681
Ensures TDS meters are calibrated properly to determine water purity and total dissolved solids.
  • For use with calibrating TDS meters
  • 1382ppm
  • 500mL
Specifications
Solution Type Calibration Standard
Type Conductivity Standard
Quantity 500 mL