missing translation for 'onlineSavingsMsg'
Learn More

TDP-43/TARDBP Antibody (DB9) - BSA Free, Novus Biologicals™

Product Code. 18690538 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18690538 0.025 mL 0.03mL
18675958 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18690538 Supplier Novus Biologicals Supplier No. NBP2503310.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TDP-43/TARDBP Monoclonal specifically detects TDP-43/TARDBP in Human samples. It is validated for ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDP-43/TARDBP
Applications ELISA, Western Blot
Classification Monoclonal
Clone DB9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000
Formulation PBS
Gene Alias ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43
Host Species Mouse
Immunogen A His-tagged recombinant protein corresponding to amino acid residues 208-414 of human TDP-43/TARDBP.
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 23435
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.