Learn More
TCR alpha/beta Antibody (Desire-1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-09020-0.025mg
 View more versions of this product
Product Code. 18622179
  • 262.00€ / 25µg
Description
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TCR alpha/beta
Monoclonal
1 mg/mL
PBS
Mouse
Protein A purified
RUO
Primary
Mouse
Purified
Flow Cytometry, Immunofluorescence, Immunoprecipitation
Desire-1
Unconjugated
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
mAb desire-1 was obtained after fusion of spleen cells from a BALB.K mouse immunised twice with KB5-C20 clone cells in RPMI 1640 medium, with non-Ig-producing myeloma cells X63-Ag8.653. Desire-1 was selected for its capacity to inhibit selectively the H-2Kb-specific cytolysis by clone KB5-C20.
0.025 mg
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
6955
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ