missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP10L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13709-1-AP-150UL
This item is not returnable.
View return policy
Description
TCP10L is a human leucine zipper protein with transcription inhibition activity.Specifications
| TCP10L | |
| Polyclonal | |
| Unconjugated | |
| TCP10L | |
| C21orf77, PRED77, t complex 10 (mouse) like, T complex protein 10A 2, TCP10 like, TCP10A 2, TCP10L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140290 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TDR4 | |
| TCP10L | |
| TCP10L Fusion Protein Ag4646 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction