missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCN1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93978-0.1ml
This item is not returnable.
View return policy
Description
TCN1 Polyclonal antibody specifically detects TCN1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TCN1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| haptocorin, TC I, TC-1, TC1haptocorrin, TCItranscobalamin-1, Transcobalamin I, transcobalamin I (vitamin B12 binding protein, R binder family) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 24-280 of human TCN1 (NP_001053.2). EICEVSEENYIRLKPLLNTMIQSNYNRGTSAVNVVLSLKLVGIQIQTLMQKMIQQIKYNVKSRLSDVSSGELALIILALGVCRNAEENLIYDYHLIDKLENKFQAEIENMEAHNGTPLTNYYQLSLDVLALCLFNGNYSTAEVVNHFTPENKNYYFGSQFSVDTGAMAVLALTCVKKSLINGQIKADEGSLKNISIYTKSLVEKILSEKKENGLIGNTFSTGEAMQALFVSSDYYNENDWNCQQTLNTVLTEISQGA | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 6947 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction