missing translation for 'onlineSavingsMsg'
Learn More

TCF2 Polyclonal Antibody, Invitrogen™

Product Code. 11521033
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11521033 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11521033 Supplier Invitrogen Supplier No. PA124459

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-24459 detects Hepatocyte Nuclear Factor 1-beta (TCF2) in Human samples. PA1-24459 has been successfully used in Western Blot procedures. PA1-24459 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human TCF2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Hepatocyte Nuclear Factor 1? (HNF-1?, Homeoprotein LFB3, Transcription factor 2, TCF2, Variant hepatic nuclear factor ) is a liver-specific factor of the homeobox-containing basic helix-turn-helix family. The TCF2 protein is believed to form heterodimers with another liver-specific member of this transcription factor family, TCF1; depending on the TCF2 isoform, the result may be to activate or inhibit transcription of target genes. Mutation of TCF2 that disrupts normal function has been identified as the cause of MODY5 (Maturity-Onset of Diabetes, Type 5). A third human transcript variant is believed to exist based on such a variant in the rat: however, to date such an mRNA species has not been isolated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Gene HNF1B
Gene Accession No. P35680
Gene Symbols HNF1B
Host Species Rabbit
Immunogen Synthetic peptide region of human TCF2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6928
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.