Learn More
TCF-12/HTF4 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-76304PCP
 View more versions of this product
Product Code. 18573620
  • 564.00€ / 0.10mL
Description
TCF-12/HTF4 Polyclonal antibody specifically detects TCF-12/HTF4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TCF-12/HTF4
Polyclonal
PBS
Rabbit
Peptide affinity purified
RUO
Primary
Human
Purified
Western Blot
PerCP
BHLHB20, bHLHb20E-box-binding protein, Class B basic helix-loop-helix protein 20, HEBDNA-binding protein HTF4, helix-loop-helix transcription factor 4, HsT17266, HTF4TCF-12, transcription factor 12, Transcription factor HTF-4
Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human TCF12. The immunogen is located within amino acids 50 - 100 of TCF12. Amino Acid Squence: QPSPSYDSSRGFTDSPHYS
0.1 mL
Immunology
6938
Store at 4°C in the dark.