missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEB3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38555-100ul
This item is not returnable.
View return policy
Description
TCEB3B Polyclonal antibody specifically detects TCEB3B in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TCEB3B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| 110kDa Elongin A, EloA2, Elongin-A2, HsT832, MGC119351, RNA polymerase II transcription factor SIII subunit A2, TCEB3Ltranscription elongation factor (SIII) elongin A2 elongin A2, Transcription elongation factor B polypeptide 3B, transcription elongation factor B polypeptide 3B (elongin A2) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 534-753 of human TCEB3B (NP_057511.2).,, Sequence:, AQVLRNNPDALSDVGEVPYWVLEPVLEGWRPDQLYRRKKDNHALVRETDELRRNHCFQDFKEEKPQENKTWREQYLRLPDAPEQRLRVMTTNIRSARGNNPNGREAKMICFKSVAKTPYDTSRRQEKSAGDADPENGEIKPASKPAGSSHTPSSQSSSGGGRDSSSSILRWLPEKRANPCLSSSNEHAAPAAKTRKQAAKKVAPLMAKAIRDYKRRFSRR | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51224 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction