missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TC-PTP/PTPN2 Antibody, Novus Biologicals™

Product Code. 30045913 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30045913 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30045913 Supplier Novus Biologicals™ Supplier No. NBP329575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TC-PTP/PTPN2 Polyclonal antibody specifically detects TC-PTP/PTPN2 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TC-PTP/PTPN2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias protein tyrosine phosphatase, non-receptor type 2, PTPTPTN2, T-cell protein tyrosine phosphatase, T-cell protein-tyrosine phosphatase, TCELLPTP, TC-PTP, TCPTPEC 3.1.3.48, tyrosine-protein phosphatase non-receptor type 2
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5771
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.