Learn More
TBX20 Antibody (OTI5D4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44934-100ul
This item is not returnable.
View return policy
Description
TBX20 Monoclonal antibody specifically detects TBX20 in Human,Mouse samples. It is validated for Western Blot
Specifications
| TBX20 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5D4 | |
| Western Blot 1:2000 | |
| ASD4, T-box 20, T-box protein 20, T-box transcription factor TBX20 | |
| Human recombinant protein fragment corresponding to amino acids 1-340 of human TBX20(NP_001071121) produced in E.coli. | |
| 100 μL | |
| Angiogenesis, Cardiovascular Biology | |
| 57057 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |