missing translation for 'onlineSavingsMsg'
Learn More

TBP like protein TLP Antibody, Novus Biologicals™

Product Code. 18028294 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18028294 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18028294 Supplier Novus Biologicals Supplier No. NB1001101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

TBP like protein TLP Polyclonal specifically detects TBP like protein TLP in Human samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBP like protein TLP
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_004856.
Gene Alias 21 kDa TBP-like protein, STUD21-kDA TBP-like protein, TATA box binding protein-related factor 2, TATA box-binding protein-like protein 1, TATA box-binding protein-related factor 2, TBP-like 1, TBP-like factor, TBP-like protein 1, TBP-related factor 2, TBP-related protein, TLFMGC:8389, TLP21, TLPMGC:9620, TRF2Second TBP of unique DNA protein, TRP
Gene Symbols TBPL1
Host Species Goat
Immunogen Peptide with sequence C-QIYPFVFESRKEIL corresponding to C-Terminus according to NP_004856.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9519
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.