missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38611-20ul
This item is not returnable.
View return policy
Description
TBC1D1 Polyclonal antibody specifically detects TBC1D1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TBC1D1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| KIAA1108TBC, TBC1, TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1, TBC1 domain family member 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1069-1168 of human TBC1D1 (NP_055988.2).,, Sequence:, IDSSPLSDNQRMDKLEKTNSSLRKQNLDLLEQLQVANGRIQSLEATIEKLLSSESKLKQAMLTLELERSALLQTVEELRRRSAEPSDREPECTQPEPTGD | |
| 20 μL | |
| Signal Transduction | |
| 23216 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction