Learn More
TB Solution-II, Thermo Scientific™
Product Code. 15412879
Click to view available options
Quantity:
125 mL
250 mL
Unit Size:
125mL
250mL
Product Code. 15412879
Brand: Thermo Scientific Alfa Aesar J60028.=E
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For transformation of competent cells
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material TB Solution-II
Color Colorless
Quantity 125 mL
Physical Form Liquid