missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau, p Ser396 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-82243
This item is not returnable.
View return policy
Description
Tau Polyclonal specifically detects Tau in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Tau (p Ser396) | |
| Polyclonal | |
| Unconjugated | |
| PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide | |
| DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects endogenous levels of Tau only when phosphorylated at serine396. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:100 | |
| P10636 | |
| MAPT | |
| This Tau [p Ser396] Antibody was produced against synthesized phosphopeptide derived from human Tau around the phosphorylation site of serine 396 (Y-K-SP-P-V). | |
| 0.05 mL | |
| Alzheimers Research, Apoptosis, Cytokine Research, Cytoskeleton Markers, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers | |
| 4137 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction