missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-82247-0.025ML
This item is not returnable.
View return policy
Description
Tau Polyclonal specifically detects Tau in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Tau | |
| Polyclonal | |
| Unconjugated | |
| phosphate buffered saline (without Mg2+and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
| DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tau (specific to a.a. 231) detects endogenous levels of total Tau protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| P10636 | |
| MAPT | |
| The antiserum was produced against synthesized non-phosphopeptide derived from human Tau around aa. 231 (V-R-T-P-P) | |
| 0.025 mL | |
| Alzheimers Research, Apoptosis, Cytokine Research, Cytoskeleton Markers, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Stem Cell Markers | |
| 4137 | |
| Mouse, Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction