Learn More
TATA binding protein TBP Antibody (SR1605), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21665-100ul
This item is not returnable.
View return policy
Description
TATA binding protein TBP Monoclonal antibody specifically detects TATA binding protein TBP in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TATA binding protein TBP | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1605 | |
| Western Blot 1:1000-1:2000 | |
| GTF2D, GTF2D1, HDL4, MGC126054, MGC126055, SCA17, TATA box binding protein, TATA sequence-binding protein, TATA-binding factor, TATA-box binding protein N-terminal domain, TATA-box factor, TATA-box-binding protein, TF2D, TFIIDMGC117320, Transcription initiation factor TFIID TBP subunit | |
| A synthesized peptide derived from human TATA binding protein TBP (Uniprot #: P20226) | |
| 100 μg | |
| Transcription Factors and Regulators | |
| 6908 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur