Learn More
Tashiro's indicator solution, In ethanol, Honeywell Fluka™
Product Code. 15665110 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
30 mL
250 mL
1 L
1.5 L
6 x 250 mL
6 x 1 L
Unit Size:
1 L
1.5 L
250mL
30mL
Pack of 6
Product Code. 15665110
Brand: Honeywell Fluka 360836X250ML
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
In ethanol
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tashiro’s indicator solution
Quantity 6 x 250 mL
Flash Point 12°C (53.6°F)