missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ GAPDH Assay, JUN™ dye/QSY™ probe
GREENER_CHOICE

Product Code. 15691802 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
250 reactions
Unit Size:
250 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15691802 250 reactions 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15691802 Supplier Applied Biosystems™ Supplier No. 4485712

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides a pre-formulated assay for quantitating human GAPDH.

This assay enables relative gene expression quantification in cDNA samples when used with other gene expression assays. The assay consists of a JUN™ dye-labeled QSY™ probe plus sequence-specific forward and reverse primers, and it can be used for multiplex or singleplex PCR reactions.

Save time in assay design, formulation, and testing by using the TaqMan™ GAPDH Assay as your control when quantifying gene expression. This pre-designed probe and primer set enables you to normalize the amount of sample RNA or DNA in a reaction when used with a control sample. Or use it for relative gene expression quantification in cDNA samples when used with other gene expression assays.

The JUN™ dye is optimized for use with the 4th filter of our QuantStudio™, ViiA™ 7, and 7500 series real-time PCR instruments. Please note that master mix containing ROX™ dye cannot be used with this assay (with JUN™ dye) due to an overlap of spectra. TaqMan™ Multiplex Master Mix with MUSTANG PURPLE™ dye is recommended.

  • Eliminate assay design time by using pre-designed primers and pre-designed JUN™ dye-labeled TaqMan™ probe
  • Minimize experimental optimization time
  • Use in conjunction with FAM™ or VIC™ dye-labeled predesigned gene expression assays
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_002046.3
Content And Storage Primer-Probe Set(s)
Detection Method Primer-probe
Format Tube
PCR Method qPCR
Gene Symbol GAPDH
Internal Probe Modification QSY™ Non-Fluorescent Quencher (3')
Label or Dye JUN
Product Line TaqMan
Product Type Gapdh Assay
Quantity 250 reactions
Species Human
Sufficient For 250 Reactions
Target Glyceraldehyde-3-phosphate dehydrogenase
For Use With (Application) Gene Expression
No. of Reactions 250 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.