missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Custom TaqMan™ Array Card is a 384-well microfluidic card designed to perform 384 simultaneous real-time PCR reactions without using liquid-handling robots or multichannel pipettors. Select target assays from thousands of inventoried TaqMan™ Gene Expression Assays.
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C). |
| Format | 96A |
| Product Line | TaqMan |
| Product Type | Custom Gene Expression TaqMan™ Array Cards |
| Quantity | 1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?