missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Arbovirus Triplex Kit (ZIKV/DENV/CHIKV), 0.1 mL

Product Code. 15689623
Change view
Click to view available options
Quantity:
96 reactions, 0.1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15689623 96 reactions, 0.1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15689623 Supplier Applied Biosystems™ Supplier No. A31747

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A simple solution for dengue, chikungunya, and Zika virus RNA detection: just add RNA.

The TaqMan Arbovirus Triplex Kit enables the simultaneous detection and differentiation of RNA from dengue virus (DENV), chikungunya virus (CHIKV), and Zika virus (ZIKV) in urine and serum. It is a lyophilized, multiplex, one-step real-time PCR Applied Biosystems TaqMan assay for the molecular detection of dengue, chikungunya, and Zika RNA. The kit is optimized for use with the Applied Biosystems QuantStudio 5 Real-Time PCR System, but is also suitable for the 7500, 7500 Fast, and other QuantStudio systems. It includes a primer-probe set for human PPIA (cyclophilin A) as an endogenous extraction, RT-PCR and PCR amplification control. The kit is compatible with RNA extracted from human urine or serum using the MagMAX Pathogen RNA/DNA Kit. The Zika virus sequence used to develop these primers and probes is from a 2014 research paper by Pyke et al (1).

Features of the kit:
• Lyophilized with one-step RT-PCR master mix; simply add RNA
• Simple workflow for detection of Zika, Dengue, and Chikungunya virus RNA from urine or serum
• Easy to use and environmentally friendly. No cold chain management required; shipped and stored at room temperature.

Contents:
• Twelve 8-well strips tubes for 96 total reactions per kit
• 0.1-mL tube format, compatible with 96-well fast block
• Primer-probe detection with 4 probe reporter dyes:
   --FAM-Zika
   --VIC-Dengue
   --ABY-Chikungunya
   --JUN-PPIA cyclophilin

1. Pyke AT, Daly MT, Cameron JN, Moore PR, Taylor CT, Hewitson GR, Humphreys JL, Gair R. Imported zika virus infection from the cook islands into australia, 2014. PLoS Curr. 2014 Jun 2;6.

Order Info

Ambient Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 12 x 8-well strip tubes, stored at room temperature
Detection Method Primer-probe
Form Lyophilized
Format Tube
GC-Rich PCR Performance Low
PCR Method 1-step RT-qPCR
Reaction Speed Fast or Standard
Disease Chikungunya, Dengue, Zika
Product Line TaqMan
Product Type Zika Virus Triplex Kit
Quantity 96 reactions, 0.1 mL
Shipping Condition Room Temperature
Sufficient For 96 Reactions
For Use With (Application) Pathogen Detection
No. of Reactions 96 Reactions
Volume 0.1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.