missing translation for 'onlineSavingsMsg'
Learn More

TAPP1/PLEKHA1 Antibody (SR1740), Novus Biologicals™

Product Code. 18634594 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18634594 50 μg 50µL
18653694 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18634594 Supplier Novus Biologicals Supplier No. NBP32250650ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TAPP1/PLEKHA1 Monoclonal antibody specifically detects TAPP1/PLEKHA1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAPP1/PLEKHA1
Applications Western Blot
Classification Monoclonal
Clone SR1740
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias family A (phosphoinositide bindingspecific) member 1, PH domain-containing family A member 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAPP1/PLEKHA1 (Uniprot #: Q9HB21)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59338
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.