missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD160 Monoclonal Antibody (342705)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561356

Product Code. 30533978

  • 429.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 is expressed at the cell surface as a tightly disulfide-linked multimer. It is involved in cell proliferation, cell surface receptor signaling pathways, cellular defense response, and regulation of immune response.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD160
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative
O88875
CD160
Mouse myeloma cell line NS0-derived recombinant mouse CD160, Gly28-Ser160.
100 μg
Primary
Mouse
Antibody
IgG2a
Immunocytochemistry
342705
Unconjugated
CD160
AU045688; BY55; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 molecule; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell BY55; natural killer cell receptor; natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member; NK1; NK28; RGD1563598
Rat
Protein A/G
RUO
54215
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.