missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAP Tag Antibody (22F12-1E3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54891-25ug
This item is not returnable.
View return policy
Description
TAP Tag Monoclonal specifically detects TAP Tag in Human, Non-species specific samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TAP Tag | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 - 1:2000 | |
| tandem affinity purification, tandem affinity purification tag, TAP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Immunofluorescence | |
| 22F12-1E3 | |
| Unconjugated | |
| PBS, 1.0 mg/mL BSA and 30% Glycerol with 0.05% Sodium Azide | |
| CDK10 | |
| KLH conjugated peptide representing the C-terminus of the TAP construct after TEV cleavage | |
| 25 μg | |
| Cell Cycle and Replication | |
| 8558 | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction