missing translation for 'onlineSavingsMsg'
Learn More

TAFA3/FAM19A3 Antibody (460904), Allophycocyanin, Novus Biologicals™

Product Code. 18638883 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18638883 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18638883 Supplier Novus Biologicals Supplier No. FAB4677A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TAFA3/FAM19A3 Monoclonal antibody specifically detects TAFA3/FAM19A3 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAFA3/FAM19A3
Applications Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 460904
Conjugate APC
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Chemokine-like protein TAFA-3, family with sequence similarity 19 (chemokine (C-C motif)-like), member A3, MGC138473, TAFA3, TAFA-3
Host Species Mouse
Immunogen E. coli-derived recombinant human TAFA3/FAM19A3, Ala31-Arg133, Accession # Q7Z5A8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284467
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.