missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF3B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
450.00€ - 708.00€
Specifications
| Antigen | TAF3B1 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617484
|
Novus Biologicals
NBP3-21302-100ul |
100 ul |
708.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608604
|
Novus Biologicals
NBP3-21302-25ul |
25 μg |
450.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAF3B1 Polyclonal antibody specifically detects TAF3B1 in Human samples. It is validated for ImmunofluorescenceSpecifications
| TAF3B1 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB, DKFZp686K0831, RNA polymerase III, GTF3B subunit 1, TAF3B1, TFIIIB150, TFIIIB90, TFNR, transcription factor-like nuclear regulator | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ALRETVRLDTSPKEMVPAEINTKEMQSDLKETGRRAISPREKILDVIDDTIEMETGLKAMGREICLREKTPEVIDATEEIDKDLEEAGR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 55814 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title