missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford TAE
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
10X solution contains:
TRIS-Base: 0.4 M
Acetic Acid: 0.2 M
EDTA: 10 mM
pH (10X Solution (25°C): 8.2-8.4
Specifications
Specifications
| CAS | 77-86-1, 64-19-7, 6381-92-6, 7732-18-5 |
| Quantity | 1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction