Learn More
Thermo Scientific Chemicals TAE (10X), TRIS + acetate + EDTA
Description
TAE is used for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis, used at various concentrations to study the mobility of DNA in solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- TAE (10X), TRIS + acetate + EDTA allows nucleic acid separation
Application
- This buffer has been used to prepare agarose and polyacrylamide gels for electrophoresis of nucleic acids
- It is used as a running and gel preparation buffer, and it is also used for non-denaturing RNA agarose gel electrophoresis
Specifications
Specifications
| Chemical Name or Material | TAE (10X) |
| Physical Form | Liquid |
| Quantity | 1 L |
| MDL Number | MFCD00236357 |
| Packaging | Plastic bottle |
| Solubility Information | Fully miscible. |