Learn More
Tadalafil, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Tadalafil |
| CAS | 171596-29-5 |
| Analyte or Component Names | Tadalafil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H19 N3 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (6R,12aR)-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-methylpyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, (6R-trans)-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-methylpyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, (6R,12aR)-2,3,6,7,12,12a-hexahydro-2-methyl-6-(3,4-methylenedioxyphenyl)pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, (6R,12aR)-6-(1,3-benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-methylpyrazino [1',2':1,6]pyrido[3,4-b]indole-1,4-dione, Cialis, GF 196960, IC 351, ICOS 351, Tadalafil, Tildenafil, UK 336017 |
| InChI Formula | InChI=1S/C22H19N3O4/c1-24-10-19(26)25-16(22(24)27)9-14-13-4-2-3-5-15(13)23-20(14)21(25)12-6-7-17-18(8-12)29-11-28-17/h2-8,16,21,23H,9-11H2,1H3/t16-,21-/m1/s1 |
| Solution Type | Neat |
| SMILES | CN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O |
| Show More |