missing translation for 'onlineSavingsMsg'
Learn More

Tadalafil Hydroxypiperidone, TRC

Product Code. 30138642
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138642 50 mg 50mg
30135252 5 mg 5mg
30012030 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138642 Supplier Toronto Research Chemicals Supplier No. TRCT00452550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1978322-99-4
Molecular Formula C22H19N3O6
Molecular Weight (g/mol) 421.4
Synonym Tadalafil EP Impurity E;
IUPAC Name benzyl (2R,3R,5R)-5-[(2R)-2-formamido-4-methylpentanoyl]oxy-2-hexyl-3-phenylmethoxyhexadecanoate
SMILES O=C1N(C)CC(N([C@@]1(C2C3(O)NC4=C(C=CC=C4)C2=O)[H])[C@]3([H])C5=CC=C(OCO6)C6=C5)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C43H67NO6/c1-5-7-9-11-12-13-14-15-22-28-38(50-43(47)40(44-34-45)30-35(3)4)31-41(48-32-36-24-18-16-19-25-36)39(29-23-10-8-6-2)42(46)49-33-37-26-20-17-21-27-37/h16-21,24-27,34-35,38-41H,5-15,22-23,28-33H2,1-4H3,(H,44,45)/t38-,39-,40-,41-/m1/s1
Formula Weight 693.4968
Chemical Name or Material Tadalafil Hydroxypiperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.