CAS RN 1978322-99-4
CAS RN 1978322-99-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tadalafil Hydroxypiperidone, TRC
CAS: 1978322-99-4 Molecular Formula: C22H19N3O6 Molecular Weight (g/mol): 421.4 Synonym: Tadalafil EP Impurity E; IUPAC Name: benzyl (2R,3R,5R)-5-[(2R)-2-formamido-4-methylpentanoyl]oxy-2-hexyl-3-phenylmethoxyhexadecanoate SMILES: O=C1N(C)CC(N([C@@]1(C2C3(O)NC4=C(C=CC=C4)C2=O)[H])[C@]3([H])C5=CC=C(OCO6)C6=C5)=O