missing translation for 'onlineSavingsMsg'
Learn More

Tacstd1 Mouse anti-Rat, Clone: Epcam/1158, Abnova™

Product Code. 16062516
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16062516 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16062516 Supplier Abnova Supplier No. MAB14196.500uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant rat Tacstd1.

  • High specificity
  • High reactivity

Specifications

Antigen Tacstd1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone Epcam/1158
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant rat Tacstd1.
Dilution Flow Cytometry (5-10 uL/million cells in 0.1 mL) Immunofluorescence (1:50-1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) Western Blot (1:100-1:200) The optimal working dilution should be determined by
Formulation In bioreactor concentrate (0.05% sodium azide).
Gene Accession No. O55159
Gene Alias Egp314
Gene Symbols Tacstd1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length rat Tacstd1.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171577
Target Species Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.