missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tacrine Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Tacrine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Tacrine Hydrochloride |
| CAS | 1684-40-8 |
| Analyte or Component Names | Tacrine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H14 N2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 9-Acridinamine, 1,2,3,4-tetrahydro-, hydrochloride (1:1), 9-Acridinamine, 1,2,3,4-tetrahydro-, monohydrochloride (9CI), Acridine, 9-amino-1,2,3,4-tetrahydro-, hydrochloride (7CI), Acridine, 9-amino-1,2,3,4-tetrahydro-, monohydrochloride (8CI), 1,2,3,4-Tetrahydro-9-aminoacridine hydrochloride, 5,6,7,8-Tetrahydroacridin-9-amine hydrochloride, 9-Amino-1,2,3,4-Tetrahydroacridine hydrochloride, CI 970, Cognex, Hydroaminacrine, Romotal, THA, Tacrin hydrochloride, Tacrine hydrochloride, Tenakrin |
| InChI Formula | InChI=1S/C13H14N2.ClH/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13;/h1,3,5,7H,2,4,6,8H2,(H2,14,15);1H |
| SMILES | Cl.Nc1c2CCCCc2nc3ccccc13 |
| IUPAC Name | 1,2,3,4-tetrahydroacridin-9-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction