Learn More
Prestige Medical T-Classic Media Autoclave
Product Code. 15802635
Change view
Click to view available options
Capacity (Metric):
12 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15802635 12 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15802635 Supplier Prestige Medical Supplier No. 210048XP
  • ONLINE EXCLUSIVE PRICE
    1816.50€
    1587.00€ / Pack of 1
    Save 229.50€
    12% Off
 Restricted Item
Estimated Shipment: 23-12-2026
T-Classic Media Autoclave, 12 L benchtop autoclave, 121°C cycle, with european plug.
T-Classic Media Autoclave, 12 L benchtop autoclave, 121°C cylce, european plug
  • Lightweight and portable
  • 12 liter capacity
  • Sterilizes at 121°C
The Classic is reliable and trusted worldwide, designed to give you the most cost effective sterilization solution. It is simplicity itself to use, fully automatic with a simple, one touch operation and guaranteed to meet your needs.
Specifications
Capacity (English) 2.6 gal.
Capacity (Metric) 12 L
Depth (Metric) Chamber 270 mm
Diameter (English) Chamber 8.4 in.
Depth (English) Chamber 10.8 in.
Diameter (Metric) Chamber 210 mm
Length (English) 13.6 in.
Length (Metric) 340 mm
Product Type Autoclave
Temperature (English) 249.8°F
Temperature (Metric) 121°C