missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T Cell Receptor Gamma/Delta Hamster, anti-Mouse, FITC, Clone: GL3, STEMCELL Technologies
Hamster Monoclonal Antibody
172.80€ - 373.95€
Specifications
Antigen | T Cell Receptor Gamma/Delta |
---|---|
Clone | GL3 |
Applications | Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Blocking Assay |
Classification | Monoclonal |
Conjugate | FITC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17139031
|
Stemcell Technologies
60104FI.1 |
50 μg |
172.80€
50µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
17129031
|
Stemcell Technologies
60104FI |
200 μg |
373.95€
200µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
- The GL3 antibody reacts with the δ chain of the murine T cell receptor γ/δ (TCRγ/δ or TCR gamma/delta), a subtype of the TCR involved in the recognition of both peptide and lipid antigens
- TCRγ/δ comprises a heterodimer (∼ 80 kDa in humans) of disulfide-linked γ and δ subunits that associates with CD3 on the cell surface
- It is a member of the immunoglobulin superfamily
- TCRγ/δ is expressed on a subpopulation of T cells in the circulation but may be found on up to 50% of the T cells in epithelial cell-rich tissues
- These cells have roles in oral and tumor-associated tolerance as well as autoimmune disease and have been described as a link between the adaptive and innate immune responses
- Once activated, they secrete effector cytokines in a subtype- and context-specific manner
- Most γ/δ T cells are CD4-/CD8-, though some express CD8
- The GL3 antibody recognizes an epitope in the constant region of the δ chain and can reportedly activate TCRγ/δ+ cells
Specifications
T Cell Receptor Gamma/Delta | |
Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Blocking Assay | |
FITC | |
Mouse | |
Primary |
GL3 | |
Monoclonal | |
Hamster | |
IgG2 κ |