Learn More
T Cell Receptor Gamma/Delta Hamster, anti-Mouse, FITC, Clone: GL3, STEMCELL Technologies
Hamster Monoclonal Antibody
Brand:  Stemcell Technologies 60104FI
Additional Details : Weight : 0.00110kg
 View more versions of this product
Product Code. 17129031
  • 373.95€ / 200µg
Estimated Shipment: 24-06-2024
Description
  • The GL3 antibody reacts with the δ chain of the murine T cell receptor γ/δ (TCRγ/δ or TCR gamma/delta), a subtype of the TCR involved in the recognition of both peptide and lipid antigens
  • TCRγ/δ comprises a heterodimer (∼ 80 kDa in humans) of disulfide-linked γ and δ subunits that associates with CD3 on the cell surface
  • It is a member of the immunoglobulin superfamily
  • TCRγ/δ is expressed on a subpopulation of T cells in the circulation but may be found on up to 50% of the T cells in epithelial cell-rich tissues
  • These cells have roles in oral and tumor-associated tolerance as well as autoimmune disease and have been described as a link between the adaptive and innate immune responses
  • Once activated, they secrete effector cytokines in a subtype- and context-specific manner
  • Most γ/δ T cells are CD4-/CD8-, though some express CD8
  • The GL3 antibody recognizes an epitope in the constant region of the δ chain and can reportedly activate TCRγ/δ+ cells
Specifications
Specifications
T Cell Receptor Gamma/Delta
Monoclonal
FITC
200 μg
Mouse
T Cells, T Cells, Other Subsets
Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Blocking Assay
GL3
Hamster
Primary
IgG2 κ