missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T-bet/TBX21 Antibody (4B10), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43298PECY55
This item is not returnable.
View return policy
Description
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Flow Cytometry.
Specifications
| T-bet/TBX21 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| TBX21 | |
| The immunogen for this antibody was TBet. | |
| 0.1 mL | |
| 30009.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 4B10 | |
| Flow Cytometry | |
| T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Human, Mouse, Rhesus Macaque | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction