Learn More
SYT8 Rabbit, anti-Human, Polyclonal, Proteintech
Product Code. 18017115 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18017115 20 μL 20µL
18078105 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18017115 Supplier Proteintech Supplier No. 206711AP20UL
Rabbit Polyclonal Antibody
The SYT8 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human SYT8. This antibody recognizes human, mouse antigen. The SYT8 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Antigen SYT8
Applications Western Blot, Immunofluorescence, ELISA
Concentration 300 μg/mL
Gene Symbols Synaptotagmin 8, synaptotagmin VIII, SYT8, SytVIII
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 90019
Target Species Human, Mouse
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG