missing translation for 'onlineSavingsMsg'
Learn More

Syntaxin-BP3/Munc18c Antibody - BSA Free, Novus Biologicals™

Product Code. 18665880 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18665880 0.02 mL 0.02mL
18604261 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18665880 Supplier Novus Biologicals Supplier No. NBP2938520.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Syntaxin-BP3/Munc18c Polyclonal antibody specifically detects Syntaxin-BP3/Munc18c in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin-BP3/Munc18c
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias MUNC18-3, MUNC18C, Platelet Sec1 protein, Protein unc-18 homolog 3, Protein unc-18 homolog C, PSP, syntaxin 4 binding protein, syntaxin binding protein 3, syntaxin-binding protein 3, Unc18-3, UNC-18C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 343-592 of human STXBP3 (NP_009200.2). KQVVHLNLAEDCMNKFKLNIEKLCKTEQDLALGTDAEGQKVKDSMRVLLPVLLNKNHDNCDKIRAILLYIFSINGTTEENLDRLIQNVKIENESDMIRNWSYLGVPIVPQSQQGKPLRKDRSAEETFQLSRWTPFIKDIMEDAIDNRLDSKEWPYCSQCPAVWNGSGAVSARQKPRANYLEDRKNGSKLIVFVIGGITYSEVRCAYEVSQAHKSCEVIIGSTHVLTPKKLLDDIKMLNKPKDKVSLIKDE
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6814
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.