missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 7 Antibody (CL0257) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52893APC
This item is not returnable.
View return policy
Description
Syntaxin 7 Monoclonal antibody specifically detects Syntaxin 7 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Syntaxin 7 | |
| Monoclonal | |
| APC | |
| syntaxin 7, syntaxin-7 | |
| This antibody was developed using a recombinant protein derived from O15400, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Endosome Markers | |
| 8417 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0257 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction