missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Syntaxin 2 Monoclonal Antibody (338824)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561226

Product Code. 30533906

  • 429.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The product of this gene belongs to the syntaxin/epimorphin family of proteins. The syntaxins are a large protein family implicated in the targeting and fusion of intracellular transport vesicles. The product of this gene regulates epithelial-mesenchymal interactions and epithelial cell morphogenesis and activation. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Syntaxin 2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P32856
Stx2
E. coli-derived recombinant human Epimorphin/Syntaxin 2, Met1-Arg188.
100 μg
Primary
Human
Antibody
IgG1
Western Blot
338824
Unconjugated
Stx2
AW538950; EPIM; Epimorphin; EPM; G1-536-1; I79_024396; MGC51014; repro34; Stx2; STX2A; STX2B; STX2C; Syn-2; syntaxin 2; syntaxin-2
Rat
Protein A/G
RUO
2054
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.