missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaphilin Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76908PCP
This item is not returnable.
View return policy
Description
Syntaphilin Polyclonal antibody specifically detects Syntaphilin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Syntaphilin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| PerCP | |
| bA314N13.5, KIAA0374, MGC46096, syntaphilin | |
| Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human Syntaphilin. The immunogen is located within amino acids 80 - 130 of Syntaphilin. Amino Acid Squence: KARLKDTQDRLQDRDTEI | |
| 0.1 mL | |
| Neuroscience | |
| 9751 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction