missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaphilin Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
575.00€
Specifications
| Antigen | Syntaphilin |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Form | Purified |
Description
Syntaphilin Polyclonal antibody specifically detects Syntaphilin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Syntaphilin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| bA314N13.5, KIAA0374, MGC46096, syntaphilin | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the center of human Syntaphilin. The immunogen is located within amino acids 350 - 400 of Syntaphilin. Amino Acid Squence: VGDELEAPEPITRGPT | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| mFluor Violet 450 SE | |
| Rabbit | |
| Neuroscience | |
| 50mM Sodium Borate | |
| 9751 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title