missing translation for 'onlineSavingsMsg'
Learn More

SYNM Monoclonal Antibody (OTI2F4), TrueMAB™, OriGene

Product Code. p-200013589
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17546893 30 μL 30µL
17526893 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17546893 Supplier Origene Technologies Supplier No. TA806626S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SYNM Monoclonal antibody specifically detects SYNM in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SYNM
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Bcl2
Gene Accession No. P10415
Gene Alias 4930412K21Rik; AI852401; Desmuslin; DMN; E130104F11; KIAA0353; SYN; synemin; synemin alpha; synemin beta; synemin, intermediate filament protein; synm
Host Species Mouse
Immunogen Full length human recombinant protein of human BCL2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 596
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.